7-Bromo[1,2,4]triazolo[1,5-a]pyridine
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR46017-500MGEA
408
EUR
APOSOR46017-500MG
7-Bromo[1,2,4]triazolo[1,5-a]pyridine
7-Bromo[1,2,4]triazolo[1,5-a]pyridine
Formule:
C₆H₄BrN₃ MW: 198,0201 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD10699200 CAS-nummer: 1053655-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...