trans-4-Dimethylaminocrotonic acid hydrochloride

Leverancier: Apollo Scientific

APOSOR471113-25GEA 98.9 EUR
APOSOR471113-25G
trans-4-Dimethylaminocrotonic acid hydrochloride
trans-4-Dimethylaminocrotonic acid hydrochloride
Formule: C₆H₁₂ClNO₂
MW: 165,62 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03695466
CAS-nummer: 848133-35-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR