2-Aminobenzo[d]thiazole-7-carboxylic acid 97%

Leverancier: Apollo Scientific

APOSOR480523-250MGEA 125 EUR
APOSOR480523-250MG
2-Aminobenzo[d]thiazole-7-carboxylic acid 97%
2-Aminobenzo[d]thiazole-7-carboxylic acid
Formule: C₈H₆N₂O₂S
MW: 194,2104 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD13195418
CAS-nummer: 71224-95-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR