1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 95%

Leverancier: Apollo Scientific

APOSOR480726-10GEA 69.8 EUR
APOSOR480726-10G
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 95%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
Formule: C₁₁H₁₂O₂
MW: 176,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02179127
CAS-nummer: 1914-65-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR