Methyl-2-piperidinecarboxylate 97%

Leverancier: Apollo Scientific

APOSOR480734-25GEA 62.6 EUR
APOSOR480734-25G
Methyl-2-piperidinecarboxylate 97%
Methyl-2-piperidinecarboxylate
Formule: C₇H₁₃NO₂
MW: 143,1836 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07364347
CAS-nummer: 41994-45-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR