1-Boc-piperidine-3-carboxylic acid

Leverancier: Apollo Scientific

Waarschuwing

APOSOR5409-5GEA 14.7 EUR
APOSOR5409-5G
1-Boc-piperidine-3-carboxylic acid
1-Boc-piperidine-3-carboxylic acid
Formule: C₁₁H₁₉NO₄
MW: 229,3 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00673775
CAS-nummer: 84358-12-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR