3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid 98%

Leverancier: Apollo Scientific

APOSOR5815-1GEA 302 EUR
APOSOR5815-1G
3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid 98%
3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid
Formule: C₁₂H₁₅BClNO₃
MW: 267,52 g/mol
Smeltpunt: 160…163 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07363775
CAS-nummer: 850589-50-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR