(±)-δ-Decalactone 95+%
Leverancier: Apollo Scientific
APOSOR6014-100GEA
69.8
EUR
APOSOR6014-100G
(±)-δ-Decalactone 95+%
(±)-δ-Decalactone
Formule:
C₁₀H₁₈O₂ MW: 170,25 g/mol Kookpunt: 117…120 °C/0,02mmHg Dichtheid: 0,954 g/cm³ (25 °C) Vlampunt: >110 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00006649 CAS-nummer: 705-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...