5-Bromo-2-cyano-3-methylpyridine

Leverancier: Apollo Scientific

Waarschuwing

APOSOR61109-1GEA 39.4 EUR
APOSOR61109-1G
5-Bromo-2-cyano-3-methylpyridine
5-Bromo-2-cyano-3-methylpyridine
Formule: C₇H₅BrN₂
MW: 197,0320 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11975788
CAS-nummer: 156072-86-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR