2,4-Dimethylphenylboronic acid
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR7209-1GEA
14.7
EUR
APOSOR7209-1G
2,4-Dimethylphenylboronic acid
2,4-Dimethylphenylboronic acid
Formule:
C₈H₁₁BO₂ MW: 149,98 g/mol Smeltpunt: 192…196 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD02683101 CAS-nummer: 55499-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...