4-Hydroxycyclohexanecarbonitrile 97%
Leverancier: Apollo Scientific
APOSOR903691-1GEA
246
EUR
APOSOR903691-1G
APOSOR903691-250MG
4-Hydroxycyclohexanecarbonitrile 97%
4-Hydroxycyclohexanecarbonitrile
Formule:
C₇H₁₁NO MW: 125,1700 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD13688990 CAS-nummer: 24056-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...