4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid 95%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR9382-1GEA
263
EUR
APOSOR9382-1G
4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid 95%
4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid
Formule:
C₁₂H₁₈BNO₅S MW: 299,1500 g/mol Smeltpunt: 157…159 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD08235037 CAS-nummer: 874219-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...