2-Methyl-5-(piperidin-1-ylsulphonyl)phenylboronic acid 98%

Leverancier: Apollo Scientific

APOSOR9392-1GEA 376 EUR
APOSOR9392-1G
2-Methyl-5-(piperidin-1-ylsulphonyl)phenylboronic acid 98%
2-Methyl-5-(piperidin-1-ylsulphonyl)phenylboronic acid
Formule: C₁₂H₁₈BNO₄S
MW: 283,1500 g/mol
Smeltpunt: 132…136 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08056364
CAS-nummer: 871333-00-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR