4-Fluoroisatin 95%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSPC201235-1GEA
166
EUR
APOSPC201235-1G
4-Fluoroisatin 95%
4-Fluoroisatin
Formule:
C₈H₄FNO₂ MW: 165,1200 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD01175824 CAS-nummer: 346-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...