3,6-Dichloro-2-fluorobenzyl alcohol 98%
Leverancier: Apollo Scientific
APOSPC302496-5GEA
212
EUR
APOSPC302496-5G
3,6-Dichloro-2-fluorobenzyl alcohol 98%
3,6-Dichloro-2-fluorobenzyl alcohol
Formule:
C₇H₅Cl₂FO MW: 195,02 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD09025388 CAS-nummer: 916420-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...