4-Bromo-2-chloro-6-fluoroanisole
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSPC3746-1GEA
46.6
EUR
APOSPC3746-1G
4-Bromo-2-chloro-6-fluoroanisole
4-Bromo-2-chloro-6-fluoroanisole
Formule:
C₇H₅BrClFO MW: 239,47 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD01631477 CAS-nummer: 261762-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...