2-Fluoro-4-iodonicotinic acid

Leverancier: Apollo Scientific

Waarschuwing

APOSPC49358-1GEA 68.4 EUR
APOSPC49358-1G
2-Fluoro-4-iodonicotinic acid
2-Fluoro-4-iodonicotinic acid
Formule: C₆H₃FINO₂
MW: 267,0000 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03095310
CAS-nummer: 884494-51-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR