2-Chloro-6-fluorocinnamic acid 98%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSPC5447-5GEA
14.7
EUR
APOSPC5447-5G
2-Chloro-6-fluorocinnamic acid 98%
2-Chloro-6-fluorocinnamic acid
Formule:
C₉H₆ClFO₂ MW: 200,6 g/mol Smeltpunt: 164…172 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00051582 CAS-nummer: 392-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...