Ytterbium(III) fluoride 99,9%
Leverancier: Apollo Scientific
|
Gevaar
|
APOSPC8022-10GEA
55.3
EUR
APOSPC8022-10G
Ytterbium(III) fluoride 99,9%
Ytterbium(III) fluoride
Formule:
F₃Yb MW: 230,04 g/mol Kookpunt: 2200 °C Smeltpunt: 1157 °C Dichtheid: 8,168 g/cm³ (25 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00049615 CAS-nummer: 13760-80-0 EINECS: 237-354-2 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...