HEPES Hemisodium Salt 0,1 M Solution in Sachets
Leverancier: Apollo
APOSBI5940P-5X10PAEA 0 EUR
APOSBI5940P-5X10PA APOSBI5940P-10PACK
HEPES Hemisodium Salt 0,1 M Solution in Sachets
Buffers Biologische buffers
Pre-weighed sachet containing sufficient buffer to create 1 litre of 0.1M solution.