VF+ hygienic hand rub refill, PURELL®
Leverancier: Gojo
PURELL®
GOJO5495-02-EEUEA 106 EUR
GOJO5495-02-EEU
VF+ hygienic hand rub refill, PURELL®
Huidverzorging Zepen en huidreinigingsmiddelen
PURELL® VF+ hygienic hand rub offers unprecedented sanitising whilst preserving healthy skin. Product benefits include a short contact time and an exceptionally broad spectrum of activity, which exceeds the European standard for hand sanitisers.
  • Kills 99% of the most common germs which may be harmful
  • Can be used throughout a hospital
PURELL® VF+ hygienic hand rub reaches the highest level of virucidal activity provided by standard EN 14476 + A1. Effective against non-enveloped viruses such as: adenovirus, rotavirus and norovirus in 30 seconds; effective against poliovirus in 45 seconds. Active against coronavirus in 30 seconds according to EN 14476 + A26. Also active against vaccinia virus (MVA) in 30 seconds according to EN 14476 + A1; an activity spectrum covering all enveloped viruses (Influenza A H1N1, HIV, Hepatitis C and Hepatitis B). Suitable for places contaminated with highly resistant viruses, such as Hepatitis A.
Certificaten: Food taint test compliant: BS EN 4120:2007.