Advanced sanitising foam, PURELL®
Leverancier: Gojo
PURELL® PURELL® Advanced
GOJO5796-04-EEU00EA 95.5 EUR
GOJO5796-04-EEU00
Advanced sanitising foam, PURELL®
Huidverzorging Zepen en huidreinigingsmiddelen
Hygienic hand sanitising foam feels great to use and is clinically proven to maintain skin health.
  • Enriched with skin conditioners that help maintain skin moisture and improve overall skin feel
  • Kills 99,99% of most common germs that may be harmful
  • Alcohol formulation 70% is suitable for frequent use by hospital staff
No-rinse hygienic hand sanitising foam is recommended for use on hands that are lightly soiled, and in conjunction with a hand hygiene regime.
Certificaten: Conforms to EN 1500, virucidal according to EN 14476, bactericidal EN 13727, EN 1276, yeasticidal EN 1650, EN 13624, fungicidal EN 1650. Food taint test compliant: BS EN 4120:2007.