Natural scrub hand cleaner, GOJO®
Leverancier: Gojo
GOJO®
GOJO7335-04-EEUEA 109 EUR
GOJO7335-04-EEU
Natural scrub hand cleaner, GOJO®
Huidverzorging Zepen en huidreinigingsmiddelen
Heavy-duty hand cleaner is effective against dirt, grease, oil, carbon black, oil-based paint and tar.
  • Contains plant-based scrubbers
  • Contains plant-based glycerin
  • Dermatologically tested
  • Fresh fragrance
This 2000 ml refill is compatible with the PRO-TDX™ dispenser.
Certificaten: Ecolabel certified.