Advanced hygienic hand rub, PURELL®
Leverancier: Gojo
PURELL®
115-2992EA 73.7 EUR
115-2992 115-2991 115-2990
Advanced hygienic hand rub, PURELL®
Huidverzorging Zepen en huidreinigingsmiddelen
A hygienic, no-rinse hand rub that feels great to use and is clinically proven to maintain skin health.
  • Includes moisturisers to help keep skin healthy
  • Kills 99,99% of most common germs that may be harmful
  • 70% alcohol formulation is suited for frequent use by hospital staff
  • Specially formulated with moisturisers to leave hands feeling soft and refreshed
  • AT-A-GLANCE™ refills; monitor product level with just one look
  • SANITARY SEALED™ PET refill is easily recycled
Recommended for use on visibly clean hands and in conjunction with a hand hygiene regime. Conforms to Bactericidal according to EN 1500 (hand sanitiser), EN 12791 (surgical rub), virucidal according to EN 14476. Passed food tainting test in accordance with EN 4120:2007.