4-(Dichloromethyl)benzonitrile 95%

BLDPBD00744661-10GEA 1167 EUR
BLDPBD00744661-10G BLDPBD00744661-5G BLDPBD00744661-1G BLDPBD00744661-100 BLDPBD00744661-250
4-(Dichloromethyl)benzonitrile 95%
4-(Dichloromethyl)benzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₅Cl₂N
MW: 186,0400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00045606
CAS-nummer: 74231-65-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR