3'-Fluoroacetophenone 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD00812534-25GEA 35 EUR
BLDPBD00812534-25G BLDPBD00812534-500 BLDPBD00812534-5G BLDPBD00812534-100 BLDPBD00812534-10G
3'-Fluoroacetophenone 97%
3'-Fluoroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₇FO
MW: 138,14 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000338
CAS-nummer: 455-36-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR