4-Bromobenzylamine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD01259492-5GEA 11 EUR
BLDPBD01259492-5G BLDPBD01259492-25G
4-Bromobenzylamine 98%
4-Bromobenzylamine
Formule: C₇H₈BrN
MW: 186,05 g/mol
MDL-nummer: MFCD00047931
CAS-nummer: 3959-07-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR