1-Octacosanol >90.0%
|
|
BLDPBD01406554-100EA
285
EUR
BLDPBD01406554-100
BLDPBD01406554-5G
BLDPBD01406554-1G
BLDPBD01406554-25G
1-Octacosanol >90.0%
1-Octacosanol
Storage condition: Sealed in dry, Room Temperature
Formule:
H₃C(CH₂)₂₆CH₂OH MW: 410,77 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00044770 CAS-nummer: 557-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...