N-Carbethoxy-4-piperidone 98%

BLDPBD01412371-25GEA 8 EUR
BLDPBD01412371-25G BLDPBD01412371-500 BLDPBD01412371-100
N-Carbethoxy-4-piperidone 98%
N-Carbethoxy-4-piperidone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₈H₁₃NO₃
MW: 171,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006188
CAS-nummer: 29976-53-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR