2'-Bromo-6'-fluoroacetophenone 98+%

BLDPBD01414813-1GEA 38 EUR
BLDPBD01414813-1G BLDPBD01414813-5G BLDPBD01414813-25G BLDPBD01414813-100
2'-Bromo-6'-fluoroacetophenone 98+%
2'-Bromo-6'-fluoroacetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₆BrFO
MW: 217,04 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD13193629
CAS-nummer: 928715-37-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR