(1S,2S)-(+)-1,2-Diaminocyclohexane 98% + (99%ee)

Leverancier: BLD PHARMATECH GMBH

BLDPBD01415045-1GEA 7 EUR
BLDPBD01415045-1G BLDPBD01415045-0.0 BLDPBD01415045-5G BLDPBD01415045-25G
(1S,2S)-(+)-1,2-Diaminocyclohexane 98% + (99%ee)
(1S,2S)-(+)-1,2-Diaminocyclohexane

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₁₄N₂
MW: 114,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00062986
CAS-nummer: 21436-03-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR