1H-Indazole-4-carbaldehyde 99%

BLDPBD01422219-5GEA 435 EUR
BLDPBD01422219-5G BLDPBD01422219-250 BLDPBD01422219-1G BLDPBD01422219100M BLDPBD01422219-25G BLDPBD01422219-100
1H-Indazole-4-carbaldehyde 99%
1H-Indazole-4-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₆N₂O
MW: 146,15 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD06738279
CAS-nummer: 669050-70-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR