6,6'-Dimethyl-2,2'-bipyridyl 99%

BLDPBD01425177-25GEA 177 EUR
BLDPBD01425177-25G BLDPBD01425177-1G BLDPBD01425177-100 BLDPBD01425177-5G
6,6'-Dimethyl-2,2'-bipyridyl 99%
6,6'-Dimethyl-2,2'-bipyridyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₂H₁₂N₂
MW: 184,2371 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00059776
CAS-nummer: 4411-80-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR