4-(4-Chlorophenyl)aniline 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD0724-1GEA 222 EUR
BLDPBD0724-1G BLDPBD0724-250MG BLDPBD0724-100MG
4-(4-Chlorophenyl)aniline 98%
4-(4-Chlorophenyl)aniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₁₂H₁₀ClN
MW: 203,6700 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00477989
CAS-nummer: 135-68-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR