4-(4-Fluorobenzoyl)piperidine hydrochloride 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD0824-5GEA 22 EUR
BLDPBD0824-5G BLDPBD0824-25G BLDPBD0824-1G BLDPBD0824-10G
4-(4-Fluorobenzoyl)piperidine hydrochloride 98%
4-(4-Fluorobenzoyl)piperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₂H₁₅ClFNO
MW: 243,71 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00044912
CAS-nummer: 25519-78-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR