4-Benzoylpiperidine hydrochloride 98%
|
|
BLDPBD0853-5GEA
162
EUR
BLDPBD0853-5G
BLDPBD0853-250MG
BLDPBD0853-1G
4-Benzoylpiperidine hydrochloride 98%
4-Benzoylpiperidine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formule:
C₁₂H₁₆ClNO MW: 225,7200 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00066982 CAS-nummer: 25519-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...