3-Fluoro-4-cyanopyridine 98%

BLDPBD100164-0.025EA 0 EUR
BLDPBD100164-0.025 BLDPBD100164-100G BLDPBD100164-1G BLDPBD100164-25G BLDPBD100164-5G BLDPBD100164-10G BLDPBD100164-250MG
3-Fluoro-4-cyanopyridine 98%
3-Fluoro-4-cyanopyridine

Storage condition: Inert atmosphere, 2-8°C

Formule: C6H3FN2
MW: 122,10 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD04112524
CAS-nummer: 113770-88-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR