N(α)-Cbz-D-tryptophan 98%

BLDPBD1005-5GEA 9 EUR
BLDPBD1005-5G BLDPBD1005-1G BLDPBD1005-100G BLDPBD1005-25G
N(α)-Cbz-D-tryptophan 98%
N(α)-Cbz-D-tryptophan

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₉H₁₈N₂O₄
MW: 338,4 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00037945
CAS-nummer: 2279-15-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR