3-Fluoro-4-nitrobenzonitrile 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD10227-10GEA 35 EUR
BLDPBD10227-10G BLDPBD10227-0.025G BLDPBD10227-100G BLDPBD10227-5G BLDPBD10227-1G BLDPBD10227-25G
3-Fluoro-4-nitrobenzonitrile 98%
3-Fluoro-4-nitrobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₃FN₂O₂
MW: 166,1093 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04973417
CAS-nummer: 218632-01-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR