5-Bromobenzimidazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD102538-1GEA 9 EUR
BLDPBD102538-1G BLDPBD102538-5G BLDPBD102538-25G BLDPBD102538-100G BLDPBD102538-10G BLDPBD102538-100MG BLDPBD102538-250MG
5-Bromobenzimidazole 98%
5-Bromobenzimidazole

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₅BrN₂
MW: 197,04 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00160001
CAS-nummer: 4887-88-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR