4-Fluorophenyl isocyanate 97%

BLDPBD10306-1GEA 7 EUR
BLDPBD10306-1G BLDPBD10306-25G BLDPBD10306-5G
4-Fluorophenyl isocyanate 97%
4-Fluorophenyl isocyanate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₇H₄FNO
MW: 137,11 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00002023
CAS-nummer: 1195-45-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR