4-Chloro-1H-imidazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD103616-5GEA 235 EUR
BLDPBD103616-5G BLDPBD103616-1G BLDPBD103616-25G BLDPBD103616-10G BLDPBD103616-250MG
4-Chloro-1H-imidazole 98%
4-Chloro-1H-imidazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₃H₃ClN₂
MW: 102,52 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD09033087
CAS-nummer: 15965-31-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR