3-Fluoro-4-(trifluoromethyl)aniline 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD10369-250MGEA 7 EUR
BLDPBD10369-250MG BLDPBD10369-1G BLDPBD10369-25G BLDPBD10369-5G BLDPBD10369-10G
3-Fluoro-4-(trifluoromethyl)aniline 97%
3-Fluoro-4-(trifluoromethyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₇H₅F₄N
MW: 179,12 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00190120
CAS-nummer: 69411-68-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR