5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD103863-250MGEA 32 EUR
BLDPBD103863-250MG BLDPBD103863-5G BLDPBD103863-10G BLDPBD103863-1G
5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one 98%
5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

Storage condition: Sealed in dry, 2-8°C

Formule: C6H8N2OS
MW: 156,2100 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00079592
CAS-nummer: 34171-37-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR