(5-Fluoropyridin-2-yl)methanol 97%

BLDPBD103960-100MGEA 22 EUR
BLDPBD103960-100MG BLDPBD103960-250MG BLDPBD103960-1G BLDPBD103960-10G BLDPBD103960-5G
(5-Fluoropyridin-2-yl)methanol 97%
(5-Fluoropyridin-2-yl)methanol

Storage condition: Inert atmosphere, 2-8°C

Formule: C₆H₆FNO
MW: 127,1200 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09027049
CAS-nummer: 802325-29-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR