4-Bromo-3-fluorotoluene 98%

BLDPBD10409-5GEA 7 EUR
BLDPBD10409-5G BLDPBD10409-25G BLDPBD10409-10G
4-Bromo-3-fluorotoluene 98%
4-Bromo-3-fluorotoluene

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₆BrF
MW: 189,03 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00040940
CAS-nummer: 452-74-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR