2-Bromo-5-iodotoluene 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD10416-10GEA 17 EUR
BLDPBD10416-10G BLDPBD10416-5G BLDPBD10416-500G BLDPBD10416-25G BLDPBD10416-100G
2-Bromo-5-iodotoluene 98%
2-Bromo-5-iodotoluene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₇H₆BrI
MW: 296,93 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00070749
CAS-nummer: 202865-85-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR