tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD104513-5GEA 174 EUR
BLDPBD104513-5G BLDPBD104513-10G BLDPBD104513-1G BLDPBD104513-250MG
tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate 96%
tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₅H₁₇NO₄
MW: 275,2998 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD05864693
CAS-nummer: 324756-80-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR