5-Nitro-o-cresol 98%

BLDPBD10528-0.025GEA 0 EUR
BLDPBD10528-0.025G BLDPBD10528-25G BLDPBD10528-10G BLDPBD10528-500G BLDPBD10528-100G
5-Nitro-o-cresol 98%
5-Nitro-o-cresol

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₇NO₃
MW: 153,14 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00043909
CAS-nummer: 5428-54-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR