2-Methoxy-5-pyridinecarboxylic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD10530-500GEA 352 EUR
BLDPBD10530-500G BLDPBD10530-5G BLDPBD10530-100G BLDPBD10530-10G BLDPBD10530-25G
2-Methoxy-5-pyridinecarboxylic acid 98%
2-Methoxy-5-pyridinecarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₇NO₃
MW: 153,1400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04115721
CAS-nummer: 66572-55-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR